Assay Details
Gene Symbol: LMNA-NTRK1 fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | OM_LMNA-NTRK1_L3N11.0 | - | 49 | 100 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LMNA-NTRK1 fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | OM_LMNA-NTRK1_L3N11.0 | - | 49 | 100 |