Assay Details
| Gene Symbol: SOX6-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SOX6-BRAF_S6B9.0 | - | 69 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOX6-BRAF fusion| Gene Symbol: SOX6-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SOX6-BRAF_S6B9.0 | - | 69 | 84 | ||