Assay Details
| Gene Symbol: ATAD2-ERBB4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ATAD2-ERBB4_A8E15.0 | - | 62 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATAD2-ERBB4 fusion| Gene Symbol: ATAD2-ERBB4 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_ATAD2-ERBB4_A8E15.0 | - | 62 | 93 | ||