Assay Details
| Gene Symbol: CCDC88C-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CCDC88C-PDGFRB_C10P12.0 | - | 50 | 81 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC88C-PDGFRB fusion| Gene Symbol: CCDC88C-PDGFRB fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CCDC88C-PDGFRB_C10P12.0 | - | 50 | 81 | ||