Assay Details
| Gene Symbol: CD74-NRG1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CD74-NRG1_C8N6.0 | - | 49 | 91 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD74-NRG1 fusion| Gene Symbol: CD74-NRG1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_CD74-NRG1_C8N6.0 | - | 49 | 91 | ||