Assay Details
| Gene Symbol: EGFR-ERP44 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EGFR-ERP44_E1E5.0 | - | 143 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EGFR-ERP44 fusion| Gene Symbol: EGFR-ERP44 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_EGFR-ERP44_E1E5.0 | - | 143 | 100 | ||