Assay Details
| Gene Symbol: GRHL2-RSPO2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GRHL2-RSPO2_G8R2.0 | - | 94 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRHL2-RSPO2 fusion| Gene Symbol: GRHL2-RSPO2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GRHL2-RSPO2_G8R2.0 | - | 94 | 76 | ||