Assay Details
| Gene Symbol: MYB-ESR1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYB-ESR1_M15E8.0 | - | 62 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYB-ESR1 fusion| Gene Symbol: MYB-ESR1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_MYB-ESR1_M15E8.0 | - | 62 | 100 | ||