Assay Details
| Gene Symbol: PAPD7-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PAPD7-RAF1_P11R10.0 | - | 51 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAPD7-RAF1 fusion| Gene Symbol: PAPD7-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PAPD7-RAF1_P11R10.0 | - | 51 | 100 | ||