Assay Details
| Gene Symbol: PDZRN3-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PDZRN3-RAF1_P5R8.0 | - | 79 | 100 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDZRN3-RAF1 fusion| Gene Symbol: PDZRN3-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PDZRN3-RAF1_P5R8.0 | - | 79 | 100 | ||