Assay Details
| Gene Symbol: GFPT1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GFPT1-ALK_G18A20_1.0 | - | 120 | 125 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GFPT1-ALK fusion| Gene Symbol: GFPT1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_GFPT1-ALK_G18A20_1.0 | - | 120 | 125 | ||
Set Membership: |
|