Assay Details
| Gene Symbol: TMCO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011541180.2 | XP_011539482.1 | 1 | 88 | 62 | |
| XM_017000916.1 | XP_016856405.1 | 1 | 88 | 62 | ||
| XM_017000920.1 | XP_016856409.1 | 1 | 88 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011541180.2, XM_017000916.1, XM_017000920.1 transcripts.
Set Membership: |
|

TMCO4
Genomic Map