Assay Details
| Gene Symbol: LOC653513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017003021.1 | XP_016858510.1 | 8 | 1572 | 103 | |
| XM_017003023.1 | XP_016858512.1 | 7 | 1461 | 103 | ||
| XM_017003024.1 | XP_016858513.1 | 8 | 3574 | 103 | ||
| XM_017003025.1 | XP_016858514.1 | 8 | 3314 | 103 | ||
| XM_017003027.1 | XP_016858516.1 | 8 | 1407 | 103 | ||
| XM_017003028.1 | XP_016858517.1 | 7 | 3463 | 103 | ||
| XM_017003029.1 | XP_016858518.1 | 7 | 3203 | 103 | ||
| XM_017003030.1 | XP_016858519.1 | 7 | 1303 | 103 | ||
| XM_017003031.1 | XP_016858520.1 | 8 | 3409 | 103 | ||
| XM_017003032.1 | XP_016858521.1 | 8 | 3149 | 103 | ||
| XM_017003036.1 | XP_016858525.1 | 7 | 3305 | 103 | ||
| XM_017003037.1 | XP_016858526.1 | 7 | 3045 | 103 | ||
| XM_017003038.1 | XP_016858527.1 | 5 | 2182 | 103 | ||
More Information
Set Membership: |
|

LOC653513
Genomic Map