Assay Details
| Gene Symbol: JDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005267332.4 | XP_005267389.1 | 1 | 2275 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005267332.4 transcripts.
Set Membership: |
|

JDP2
Genomic Map