Assay Details
| Gene Symbol: FEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004111.5 | NP_004102.1 | 2 | 516 | 70 | |
| GenBank mRNA | AK312761.1 | - | 2 | 219 | 70 | |
| BC000323.2 | - | 2 | 232 | 2821792 | 70 | |
| BP244711.1 | - | 2 | 496 | 70 | ||
| BT019524.1 | - | 1 | 103 | 70 | ||
| CR536562.1 | - | 1 | 103 | 70 | ||
| L37374.1 | - | 1 | 103 | 70 | ||
| X76771.1 | - | 2 | 470 | 70 | ||
More Information
Set Membership: |
|

FEN1
Genomic Map