Assay Details
| Gene Symbol: INF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031714.3 | NP_001026884.3 | 8 | 1201 | 91 | |
| NM_022489.3 | NP_071934.3 | 8 | 1201 | 91 | ||
| XM_005268004.4 | XP_005268061.1 | 8 | 1288 | 91 | ||
| XM_005268005.4 | XP_005268062.1 | 8 | 1259 | 91 | ||
| XM_017021595.1 | XP_016877084.1 | 8 | 1259 | 91 | ||
| GenBank mRNA | BQ961756.1 | - | 5 | 487 | 91 | |
More Information
Set Membership: |
|

INF2
Genomic Map