Assay Details
| Gene Symbol: RAB27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005254576.4 | XP_005254633.1 | 1 | 1385 | 72 | |
| XM_011521855.2 | XP_011520157.1 | 1 | 1385 | 72 | ||
| GenBank mRNA | DA553342.1 | - | 1 | 44 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005254576.4, XM_011521855.2 transcripts.
Set Membership: |
|

RAB27A
Genomic Map