Assay Details
Gene Symbol: LOC100126784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015384.2 | 1 | 488 | 73 | ||
GenBank mRNA | BC029949.1 | - | 1 | 379 | 73 |
Gene Symbol: NAV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ488207.1 | - | 1 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AJ488207.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
