Assay Details
| Gene Symbol: ACTR3BP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045000.1 | 1 | 918 | 165 | ||
| GenBank mRNA | AY026352.1 | - | 1 | 608 | 165 | |
| Gene Symbol: ACTR3BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027714.1 | 1 | 623 | 165 | ||
| GenBank mRNA | AY026351.1 | - | 1 | 623 | 165 | |
| Gene Symbol: ACTR3BP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY026350.1 | - | 1 | 605 | 165 | |
More Information
Set Membership: |
|

ACTR3BP5
Genomic Map