Assay Details
Gene Symbol: LRRC37BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK124759.1 | - | 2 | 3307 | 125 | |
AL713754.1 | - | 2 | 2426 | 125 |
Gene Symbol: LOC107133515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015341.2 | 9 | 3163 | 125 |
Gene Symbol: LRRC37B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK123784.1 | - | 1 | 1026 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
