Assay Details
Gene Symbol: HSD17B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK127832.1 | - | 1 | 1638 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK127832.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
