Assay Details
| Gene Symbol: ALOX12P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002710.2 | 14 | 2444 | 101 | ||
| NR_120453.1 | 13 | 1891 | 101 | |||
| GenBank mRNA | AF020774.1 | - | 13 | 1892 | 100 | |
| AL832768.1 | - | 14 | 2161 | 101 | ||
| BC041851.1 | - | 16 | 2625 | 101 | ||
| BU689660.1 | - | 1 | 332 | 101 | ||
More Information
Set Membership: |
|

ALOX12P2
Genomic Map