Assay Details
| Gene Symbol: TRAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011525505.2 | XP_011523807.1 | 1 | 316 | 88 | |
| XM_011525506.2 | XP_011523808.1 | 1 | 314 | 88 | ||
| XM_011525507.2 | XP_011523809.1 | 1 | 310 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011525505.2, XM_011525506.2, XM_011525507.2 transcripts.
Set Membership: |
|

TRAF4
Genomic Map