Assay Details
| Gene Symbol: ZNF347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172674.1 | NP_001166145.1 | 5 | 2905 | 193 | |
| NM_001172675.1 | NP_001166146.1 | 5 | 2585 | 193 | ||
| NM_032584.2 | NP_115973.2 | 5 | 2582 | 193 | ||
| XM_005259335.4 | XP_005259392.1 | 5 | 2904 | 193 | ||
| XM_005259336.4 | XP_005259393.1 | 5 | 2694 | 193 | ||
| XM_017027384.1 | XP_016882873.1 | 5 | 2924 | 193 | ||
| XM_017027385.1 | XP_016882874.1 | 5 | 2608 | 193 | ||
| XM_017027386.1 | XP_016882875.1 | 4 | 2530 | 193 | ||
| GenBank mRNA | AK095593.1 | - | 1 | 2143 | 193 | |
| AK096623.1 | - | 5 | 2907 | 193 | ||
| AK289994.1 | - | 5 | 2569 | 193 | ||
| AL713691.1 | - | 1 | 1564 | 193 | ||
| AY029765.1 | - | 5 | 2544 | 193 | ||
More Information
Set Membership: |
|

ZNF347
Genomic Map