Assay Details
| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127147.1 | - | 1 | 224 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK127147.1 transcripts.
Set Membership: |
|

TMEM205
Genomic Map