Assay Details
| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013301.2 | NP_037433.2 | 6 | 1872 | 119 | |
| XM_005258827.2 | XP_005258884.1 | 6 | 1470 | 119 | ||
| XM_005258828.2 | XP_005258885.1 | 6 | 1491 | 119 | ||
| GenBank mRNA | AF054984.1 | - | 6 | 1425 | 119 | |
| AK097095.1 | - | 5 | 2043 | 120 | ||
| BC000412.2 | - | 5 | 1451 | 2961290 | 120 | |
| BC008956.2 | - | 5 | 1451 | 2961290 | 120 | |
| GU354321.1 | - | 6 | 1227 | 119 | ||
| GU354322.1 | - | 6 | 1283 | 119 | ||
| GU354323.1 | - | 6 | 1471 | 119 | ||
| U79303.1 | - | 6 | 1425 | 119 | ||
More Information
Set Membership: |
|

CCDC106
Genomic Map