Assay Details
| Gene Symbol: ACVR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011512108.2 | XP_011510410.1 | 1 | 1167 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011512108.2 transcripts.
Set Membership: |
|

ACVR1
Genomic Map