Assay Details
| Gene Symbol: RRAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102669.2 | NP_001096139.1 | 6 | 1757 | 103 | |
| NM_001177314.1 | NP_001170785.1 | 6 | 1602 | 103 | ||
| NM_001177315.1 | NP_001170786.1 | 6 | 1734 | 103 | ||
| NM_012250.5 | NP_036382.2 | 6 | 1908 | 103 | ||
| XM_017017363.1 | XP_016872852.1 | 6 | 1573 | 103 | ||
| GenBank mRNA | BQ020351.1 | - | 1 | 103 | ||
More Information
Set Membership: |
|

RRAS2
Genomic Map