Assay Details
| Gene Symbol: C11orf63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005271680.4 | XP_005271737.1 | 9 | 3736 | 119 | |
| XM_011543009.2 | XP_011541311.1 | 9 | 3944 | 119 | ||
More Information
Set Membership: |
|

C11orf63
Genomic Map