Assay Details
| Gene Symbol: RBM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286045.1 | NP_001272974.1 | 4 - 5 | 579 | 104 | |
| NM_001286046.1 | NP_001272975.1 | 4 - 5 | 575 | 104 | ||
| NM_001286047.1 | NP_001272976.1 | 3 - 4 | 416 | 104 | ||
| NM_001286048.1 | NP_001272977.1 | 2 - 3 | 328 | 104 | ||
| NM_016090.3 | NP_057174.1 | 4 - 5 | 576 | 104 | ||
| GenBank mRNA | AF156098.1 | - | 4 - 5 | 454 | 104 | |
| AK002015.1 | - | 4 - 5 | 492 | 104 | ||
| AK225615.1 | - | 4 - 5 | 445 | 104 | ||
| AK225667.1 | - | 4 - 5 | 470 | 104 | ||
| AK312619.1 | - | 4 - 5 | 532 | 104 | ||
| BC034381.1 | - | 4 - 5 | 443 | 5180771 | 104 | |
| BC070262.1 | - | 4 - 5 | 437 | 4619267 | 104 | |
| BQ067937.1 | - | 197 | 104 | |||
| CN308273.1 | - | 3 - 4 | 332 | 104 | ||
More Information
Set Membership: |
|

RBM7
Genomic Map