Assay Details
| Gene Symbol: ISCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030940.3 | NP_112202.2 | 4 | 1049 | 109 | |
| GenBank mRNA | AF038186.1 | - | 1 | 464 | 109 | |
| AF284752.1 | - | 4 | 1004 | 109 | ||
| AK055643.1 | - | 4 | 1012 | 109 | ||
| AK225937.1 | - | 4 | 1012 | 109 | ||
| AL834356.1 | - | 4 | 994 | 109 | ||
| BC002675.2 | - | 4 | 1003 | 3608301 | 109 | |
| BC053900.1 | - | 4 | 970 | 109 | ||
| BC071621.1 | - | 4 | 983 | 5107151 | 109 | |
More Information
Set Membership: |
|

ISCA1
Genomic Map