Assay Details
| Gene Symbol: GPCPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006723596.3 | XP_006723659.1 | 1 | 570 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006723596.3 transcripts.
Set Membership: |
|

GPCPD1
Genomic Map