Assay Details
Gene Symbol: LZTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318099.1 | NP_001305028.1 | 4 | 1394 | 92 | |
NM_001318100.1 | NP_001305029.1 | 4 | 1471 | 92 | ||
NM_001318101.1 | NP_001305030.1 | 2 | 672 | 92 | ||
NM_032429.3 | NP_115805.1 | 4 | 1501 | 92 | ||
XM_005270222.4 | XP_005270279.1 | 4 | 1733 | 92 | ||
XM_017016780.1 | XP_016872269.1 | 3 | 3862 | 92 | ||
XM_017016781.1 | XP_016872270.1 | 3 | 842 | 92 | ||
GenBank mRNA | AB058716.1 | - | 3 | 4353 | 92 | |
AK097997.1 | - | 3 | 1670 | 92 | ||
AK298208.1 | - | 4 | 1359 | 92 | ||
AL834338.1 | - | 2 | 877 | 92 | ||
AY029201.1 | - | 3 | 1215 | 92 | ||
BC005855.2 | - | 1 | 205 | 92 | ||
BC006212.2 | - | 4 | 1359 | 3161855 | 92 | |
BC058938.1 | - | 4 | 1434 | 3913470 | 92 | |
CX755354.1 | - | 2 | 257 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
