Assay Details
Gene Symbol: TEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB051442.1 | - | 1 | 5079 | 96 |
Gene Symbol: LOC100129648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK097637.1 | - | 1 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
