Assay Details
| Gene Symbol: EIF1B-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033965.1 | 2 - 3 | 539 | 81 | ||
| GenBank mRNA | AK309874.1 | - | 2 - 3 | 349 | 81 | |
| BC031275.1 | - | 2 - 3 | 302 | 81 | ||
| BC045736.1 | - | 2 - 3 | 383 | 81 | ||
More Information
Set Membership: |
|

EIF1B-AS1
Genomic Map