Assay Details
Gene Symbol: PNRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017761.3 | NP_060231.1 | 3 | 2049 | 87 | |
XM_017001691.1 | XP_016857180.1 | 3 | 1954 | 87 | ||
GenBank mRNA | AF374386.1 | - | 3 | 1968 | 87 | |
AK000319.1 | - | 2 | 1770 | 87 | ||
AK024774.1 | - | 1 | 1469 | 87 | ||
BC001959.1 | - | 3 | 2016 | 3536977 | 87 | |
BC078177.1 | - | 3 | 1996 | 6526331 | 87 | |
BC085018.1 | - | 3 | 2019 | 6650445 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
