Assay Details
| Gene Symbol: HP09053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110821.1 | 3 | 1373 | 78 | ||
| GenBank mRNA | AB593151.1 | - | 3 | 1359 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HP09053| Gene Symbol: HP09053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110821.1 | 3 | 1373 | 78 | ||
| GenBank mRNA | AB593151.1 | - | 3 | 1359 | 78 | |
Set Membership: |
|