Assay Details
| Gene Symbol: MALAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002819.3 | 1 | 327 | 99 | ||
| GenBank mRNA | EF177381.1 | - | 1 | 327 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MALAT1| Gene Symbol: MALAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002819.3 | 1 | 327 | 99 | ||
| GenBank mRNA | EF177381.1 | - | 1 | 327 | 99 | |
Set Membership: |
|