Assay Details
| Gene Symbol: LCORL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011513821.2 | XP_011512123.1 | 7 | 2000 | 122 | |
| XM_011513822.2 | XP_011512124.1 | 7 | 2000 | 122 | ||
| XM_011513823.2 | XP_011512125.1 | 6 | 1846 | 122 | ||
| XM_017007961.1 | XP_016863450.1 | 6 | 3282 | 122 | ||
| XM_017007962.1 | XP_016863451.1 | 6 | 1924 | 122 | ||
| XM_017007963.1 | XP_016863452.1 | 6 | 2227 | 122 | ||
More Information
Set Membership: |
|

LCORL
Genomic Map