Assay Details
| Gene Symbol: LOC284648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036490.1 | 1 | 73 | 82 | ||
| GenBank mRNA | AW297673.1 | - | 1 | 73 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284648| Gene Symbol: LOC284648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036490.1 | 1 | 73 | 82 | ||
| GenBank mRNA | AW297673.1 | - | 1 | 73 | 82 | |
Set Membership: |
|