Assay Details
Gene Symbol: OCLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205254.1 | NP_001192183.1 | 9 | 5314 | 63 | |
NM_001205255.1 | NP_001192184.1 | 7 | 4497 | 63 | ||
NM_002538.3 | NP_002529.1 | 9 | 5544 | 63 | ||
XM_017008913.1 | XP_016864402.1 | 8 | 5152 | 63 | ||
XM_017008914.1 | XP_016864403.1 | 8 | 5127 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
