Assay Details
| Gene Symbol: KLRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017018684.1 | XP_016874173.1 | 7 | 1502 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLRG1| Gene Symbol: KLRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017018684.1 | XP_016874173.1 | 7 | 1502 | 96 | |
Set Membership: |
|