Assay Details
Gene Symbol: LYRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020466.4 | NP_065199.1 | 3 | 904 | 132 | |
NR_028493.1 | 3 | 1063 | 132 | |||
NR_028494.1 | 3 | 1046 | 132 | |||
NR_028495.1 | 3 | 914 | 132 | |||
GenBank mRNA | AK094994.1 | - | 3 | 973 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
