Assay Details
| Gene Symbol: VNN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011408.1 | XP_016866897.1 | 9 | 2242 | 116 | |
| XM_017011409.1 | XP_016866898.1 | 9 | 2135 | 116 | ||
More Information
Set Membership: |
|

VNN2
Genomic Map