Assay Details
| Gene Symbol: LOC441239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011516745.2 | XP_011515047.1 | 3 | 1677 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC441239| Gene Symbol: LOC441239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011516745.2 | XP_011515047.1 | 3 | 1677 | 61 | |
Set Membership: |
|