Assay Details
| Gene Symbol: SBF1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027765.2 | 1 | 182 | 80 | ||
| GenBank mRNA | AW297679.1 | - | 1 | 117 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SBF1P1| Gene Symbol: SBF1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027765.2 | 1 | 182 | 80 | ||
| GenBank mRNA | AW297679.1 | - | 1 | 117 | 80 | |
Set Membership: |
|