Assay Details
| Gene Symbol: LMLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136049.2 | NP_001129521.2 | 7 - 8 | 854 | 116 | |
| NM_033029.3 | NP_149018.2 | 7 - 8 | 854 | 116 | ||
| NR_026786.1 | 7 - 8 | 733 | 116 | |||
| NR_026787.1 | 7 - 8 | 733 | 116 | |||
| XM_011513271.1 | XP_011511573.1 | 7 - 8 | 837 | 116 | ||
| GenBank mRNA | AJ312398.1 | - | 7 - 8 | 733 | 116 | |
| AJ312399.1 | - | 7 - 8 | 854 | 116 | ||
| AK299119.1 | - | 7 - 8 | 821 | 116 | ||
| AL832783.1 | - | 6 - 7 | 1327 | 116 | ||
| AM920777.1 | - | 7 - 8 | 832 | 116 | ||
| AM920778.1 | - | 7 - 8 | 676 | 116 | ||
More Information
Set Membership: |
|

LMLN
Genomic Map