Assay Details
| Gene Symbol: TTC39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017014310.1 | XP_016869799.1 | 1 - 2 | 400 | 78 | |
| XM_017014311.1 | XP_016869800.1 | 1 - 2 | 400 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017014310.1, XM_017014311.1 transcripts.
Set Membership: |
|

TTC39B
Genomic Map