Assay Details
| Gene Symbol: COL15A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011518214.2 | XP_011516516.1 | 1 | 66 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011518214.2 transcripts.
Set Membership: |
|

COL15A1
Genomic Map