Assay Details
| Gene Symbol: EHMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024757.4 | NP_079033.4 | 27 | 4687 | 120 | |
| XM_005266105.4 | XP_005266162.1 | 27 | 4730 | 120 | ||
| XM_005266110.1 | XP_005266167.1 | 26 | 4602 | 120 | ||
| XM_006717288.2 | XP_006717351.1 | 27 | 4648 | 120 | ||
| XM_011519021.2 | XP_011517323.1 | 27 | 5685 | 120 | ||
| XM_011519022.2 | XP_011517324.1 | 27 | 5140 | 120 | ||
| XM_011519023.2 | XP_011517325.1 | 27 | 5664 | 120 | ||
| XM_011519024.2 | XP_011517326.1 | 26 | 5608 | 120 | ||
| XM_011519025.1 | XP_011517327.1 | 27 | 4850 | 120 | ||
| XM_011519026.2 | XP_011517328.1 | 26 | 5543 | 120 | ||
| XM_011519029.2 | XP_011517331.1 | 19 | 3275 | 120 | ||
| XM_011519030.2 | XP_011517332.1 | 13 | 2970 | 120 | ||
| XM_011519031.1 | XP_011517333.1 | 13 | 2328 | 120 | ||
| XM_011519033.2 | XP_011517335.1 | 26 | 5522 | 120 | ||
| XM_017015134.1 | XP_016870623.1 | 27 | 5118 | 120 | ||
| XM_017015135.1 | XP_016870624.1 | 27 | 4670 | 120 | ||
| XM_017015136.1 | XP_016870625.1 | 26 | 4592 | 120 | ||
| XM_017015137.1 | XP_016870626.1 | 27 | 4829 | 120 | ||
| XM_017015138.1 | XP_016870627.1 | 26 | 4581 | 120 | ||
| GenBank mRNA | AF461894.1 | - | 4 | 2374 | 120 | |
| BC011608.2 | - | 17 | 2904 | 120 | ||
| CA426038.1 | - | 1 | 120 | |||
More Information
Set Membership: |
|

EHMT1
Genomic Map